Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fenobam, Tocris Bioscience™
Shop All R&D Systems Products
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective non-competitive mGlu5 negative allosteric modulator (NAM) that also displays inverse agonist properties; blocks mGlu5 constitutive activity in vitro (IC50 = 87 nM). Acts at an allosteric modulatory site shared with MPEP and binds the mGlu5 receptor with Kd values of 54 and 31 nM for rat and human receptors respectively. Displays anxiolytic activity following oral administration in vivo; also exhibits analgesic properties.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 266.69 |
Percent Purity | >99% |
Chemical Name or Material | Fenobam |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction