Learn More
Corning™ Fetal Bovine Serum, 500 mL
Fetal Bovine Serum (FBS) is a time tested, universal supplement used in cell culture. It provides many compounds that have been shown to satisfy specific metabolic requirements for the culture of cells, including hormones, growth factors, cell attachment factors, pH buffer, and more.
Supplier: Corning™ 35078CV
Description
- Fetal Bovine Serum (FBS) is a time tested, universal supplement used in cell culture.
- It provides many compounds that have been shown to satisfy specific metabolic requirements for the culture of cells, including hormones, growth factors, cell attachment factors, pH buffer, protease inhibitors, vitamins, minerals, lipids, and a variety of undefined components. Serum also protects against toxic effects associated with pH change, the presence of heavy metal ions, endotoxin, and proteolytic activity.
- Sera Processing: Using industry leading techniques, blood is aseptically collected at abattoirs, centrifuged to isolate the serum and immediately frozen for testing and further processing.
- The raw sera is then true pooled and is triple 0.1 micron sterile filtered with a Sterility Assurance Level (SAL) of 10-3.
- Every lot then must pass USP <71> and EP 2.6.1 sterility testing, growth promotion testing, as well as test negative for mycoplasma and number of industry-monitored viruses.
- Authenticity of the serum is verified using industry leading measurements of Gamma-Glutamyl Transferase and Corning's high internal standard of ≤ 10 IU/L.
- Additional testing includes endotoxin, hemoglobin, pH, osmolality, total protein, and a biochemical profile that contains various metals, proteins, hormones and other metabolites.
- Origin Statement: Corning's premium FBS is only collected from government approved abattoirs in the United States, Australia, and New Zealand. All fetal bovine serum is derived from fetuses collected from healthy bovine cows and heifers that have passed ante- and post-mortem inspection and were found free of contagious diseases.
- Corning's regular FBS is derived from government registered facilities in North, Central, and South America and outside the United States.
- All fetal bovine serum is derived from fetuses collected from healthy bovine cows and heifers that have passed ante- and post-mortem inspection and were found free of contagious diseases.
- The material was exported from the country of origin accompanied with a Veterinarian Health Certificate.
Specifications
Fetal | |
Liquid | |
Fetal Bovine Serum | |
Gamma-irradiated | |
Gamma-irradiated |
New Zealand | |
Bottle | |
500 mL | |
Sterile |
Safety and Handling
ShelfLife : 60 months
Your input is important to us. Please complete this form to provide feedback related to the content on this product.