Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Fetal Bovine Serum, CELLect™ Gold
Superior for hybridoma cloning
Supplier: MP Biomedicals™ 092916849
Description
- Collected from a USDA regulated and monitored abattoir by technicians who work “onsite”
- Aseptically collected via cardiac puncture
- Supplied sterile, with no detectable mycoplasma or viral infection
Specifications
Used for cell culture and insect cell culture applications | |
3.1 to 4.4% | |
Sterile | |
USDA | |
Yes | |
27mg/dL | |
No | |
-20°C | |
Animal Serum Culture Media |
1 x 100mL | |
Heat Inactivated | |
Pale to Golden Brown | |
<1ng/mL | |
Liquid | |
280 to 325mOsm/kg | |
6.7 to 7.8 | |
USDA Inspected |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction