Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Probes™ Fetal Hemoglobin Control Kit (FETALtrol™)
SDP

Catalog No. FH101
Click to view available options
:

FETALtrol™ is a tri-level, assayed human blood control designed to help document and monitor values obtained from test methods used to determine fetal RBCs in maternal blood samples.

FETALtrol™ is a tri-level, assayed human blood control designed to help document and monitor values obtained from test methods used to determine fetal RBCs in maternal blood samples. It monitors preparation techniques, stains, reagents, and methods of data analysis for quantitative and qualitative tests. FETALtrol™ is reagent comprising D-antigen (Rho) negative adult human erythrocytes supplemented with D-antigen (Rho) positive human cord blood erythrocytes.

Application

The laboratory determination of the level of fetal cells in maternal circulation remains an important support in the obstetrical management of women with suspected uterine trauma and in the proper dose administration of Rh immune globulin. The Trillium FETALtrol™ kit provides a means of monitoring preparation techniques, stains, reagents, and methods of data analysis for quantitative and qualitative tests.

Additional Information

Use the Spectraviewer to help you plan your experiments. Additional general information about Life Technologies flow cytometry products and support is also available.

Specifications

Content And Storage

Store in refrigerator (2°C to 8°C).

Format

Tube

For Use With (Equipment)

Microscope (KBB assay-compatible), Flow Cytometer

Type

Fetal Hemoglobin Control Kit

For Use With (Application)

Flow Cytometry, Immunohistochemistry

Quantity

50 Tests

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.