Learn More
TARGETMOL CHEMICALS INC FFN 102 MESYLATE 5MG
Supplier:  TARGETMOL CHEMICALS INC T41194
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. FFN 102 mesylate is a pH responsive fluorescent false neurotransmitter (FFN) a selective dopamine transporter (DAT) and VMAT2 substrate. Exhibits no significant binding to a panel of 38 CNS receptors including dopamine and serotonin receptors. Inhibits dopamine uptake. Excitation maxima are 340 nm at pH 5 and 370 nm at pH 7.5. Emission maximum is 435 nm at both pH 5 and 7.5.FFN 102 mesylate is a Ph-responsive fluorescent pseudoneurotransmitter (FFN) that acts as a substrate for selective dopamine transporters (DAT) and VMAT2. purity: 98%
Catalog No. 50-257-9952
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.