Learn More
Thermo Scientific Beta Cyanoethyl Phosphoramidite, dA(Bz), 98796-53-3, MFCD00036314, 2 g
Supplier:  Thermo Scientific 401159
Encompass
MF: C47 H52 N7 O7 P, MW: 857.944, Chemical Name: dA(Bz) beta CEP. The dA(Bz) Beta Cyanoethyl Phosphoramidite is used as a component of a deprotecting chemistry set including dC(Bz), dG(iBu) and T. Standard series of phosphoramidite deprotects in 6-8 hours at 55°C.
Catalog No. NC0382654
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.