Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Finasteride, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. p-7157445 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle

F1300, 98319-26-7, C23H36N2O2

Spectrum™ Chemical Finasteride, USP

Chemical Identifiers

CAS 98319-26-7
Molecular Formula C23H36N2O2
Molecular Weight (g/mol) 372.55
InChI Key DBEPLOCGEIEOCV-WSBQPABSSA-N
IUPAC Name (4aR,4bS,6aS,7S,9aS,9bS,11aR)-N-tert-butyl-4a,6a-dimethyl-2-oxo-1H,2H,4aH,4bH,5H,6H,6aH,7H,8H,9H,9aH,9bH,10H,11H,11aH-indeno[5,4-f]quinoline-7-carboxamide
SMILES CC(C)(C)NC(=O)[C@H]1CC[C@H]2[C@@H]3CC[C@H]4NC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Packaging Amber Glass Bottle
Quantity 5 g
Specific Rotation -56.0° to -60.0°
Percent Purity 98.5 to 101%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.