Learn More
Electron Microscopy Sciences Embra Finder 300 mesh Grid, Copper
Supplier:  Electron Microscopy Sciences EF300CU
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Standard 3.05 mm diameter grids which have one straight and one round cut out from the rim which assists in the orientation of the grid.
Grid: Copper
Catalog No. 50-259-14
missing translation for 'QuantityLeft'