Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Fingerless Anti-Impact Gloves with Thumb Padding & Detachable Wrist Support

Catalog No. p-7214852 Shop All Impacto Protective Products Products
Click to view available options
Size:
Small
Large
Hand:
Left
Specific
This item is not returnable. View return policy
This item is not returnable. View return policy

Detachable suede leather wrist support helps to prevent over flexion and over extension. Additional padding in the thumb webbing prevents cumulative trauma that can occur from repetitive gripping.

  • Nylon fabric ensures breathability and comfort
  • Contoured VEP 1/8 in. padding in the palm and thumb web area protects from impact
  • Fingerless style for tactile feedback and mobility
  • Suede leather cover offers abrasion protection
  • Detachable leather wrist support prevents over flexion and over extension to help prevent RSI
  • For protection from hand tools and power tools
  • Ideal for applications in construction, assembly, manufacturing, maintenance. and warehousing

Specifications

Back Material Nylon Lycra™
Cuff Material Suede Leather
Color Blue, Gray
Hand Specific
Material Suede
Product Type Anti-impact Gloves
Size Small
Finger Style Fingerless
For Use With (Application) Construction, Assembly, Manufacturing, Maintenance, and Warehousing
Glove Cuff Color Gray
Series 704-10
Length (English) 7 in.
Length (Metric) 18 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.