Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trajan™ Fixed-Needle Autosampler Syringes for Thermo Scientific Instruments

For reduced non specific sample interaction and increased analyte recovery

Supplier:  Trajan™ 0029871

 View more versions of this product

Catalog No. 03-351-114


Description

Description

  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Thermo Scientific instruments
Specifications

Specifications

23 G
0.63 mm
2 in.
10F-BT-GT-5/0.63C
10 μL
Cone
Fixed
0.025 in.
Gas chromatography
Autosampler Syringe
1 Ea.
50 mm
Thermo AS2000
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.