Learn More
White Knight™ Flame-Resistant Women's Lab Coat
Catalog No. p-7202961
Click to view available options
Size:
2X-Small
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
These flame-resistant lab coats were developed specifically to fit the female form while also providing the protection required in many of today’s high-tech laboratories and research facilities.
  • Best in class flammability protection
  • Meets NFPA 70E and HRC 1 performance standards
  • Reusable Indura™ FR fabric
  • Quick-release snap front
  • Safety sleeve cuff with tunneled elastic
  • Princess-belt on back
  • FR identification patches on sleeves and back
  • Excellent heat-resistance properties
Specifications
Size 2X-Small
Gender Unisex
Material FR Cotton
Color Blue
Closure Type Snaps
Cuff Style Elastic
Disposable Reusable
Sterility Non-sterile
No. of Pockets 3
Collar Type Lapel
Latex Free Yes
Arc Flash Rating HRC 1
Certifications/Compliance NFPA 70 E compliant
Show More Show Less
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.