Learn More
MTC Bio Flask, Erlenmeyer, 1000ml, Polycarbonate, baffled bottom w vented screw cap, Sterile, indiv wrapped, 6/CS
Supplier: MTC Bio F4063B
MTC Bio’s Erlenmeyer Shaker Flasks are made of polycarbonate, PETG, or styrene/butadiene (Jumbo Flasks only). The polypropylene screw caps are vented with 0.2µm membranes to increase air transmission and enhance cell growth. The mouth size conforms with international SBS standards, for quick, secure connection to bioreactor equipment. Polycarbonate Flasks and Jumbo Flasks have baffled (ribbed) bottoms to provide increased oxygenation and mixing efficiency, even at lower shaking speeds. All flasks, regardless of size or material are individually wrapped and gamma sterilized to SAL 10-6, and are free of RNase, DNase, pyrogen m endotoxins and human DNA.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.