Learn More
Chemglass Life Sciences Flask, Shake, 500mL, Deep Baffles, Plain Top
Supplier: Chemglass Life Sciences CLS204204
Flask, Shake, 500mL, Deep Baffles, Plain Top, Approx OD x Height (mm): 100 x 180. Extra deep baffle indents on the lower outside edge of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.