Learn More
Chemglass Life Sciences Flask, Shake, 2L, 6 Bottom Baffles, 70mm Solid Cap, Approx OD x Height (mm): 170 x 270
SDP
Supplier:  Chemglass Life Sciences CLS205406
Encompass_Preferred
Flask, Shake, 2L, 6 Bottom Baffles, 70mm Solid Cap, Approx OD x Height (mm): 170 x 270. Flask has 6 bottom baffles to maximize oxygen transfer for bacterial, yeast and other cultures that require large amounts of oxygen for optimal growth. The 70mm vented cap has a 0.20µm PVDF hydrophobic membrane to allow for sterile gas exchange while preventing contamination. A silicone liner ensures a tight seal between the cap and flask. A solid cap is available for anaerobic cultures. The flasks, cap and liners are all fully autoclavable.
Catalog No. 50-143-833
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.