Learn More
Electron Microscopy Sciences Flat Emery Stick 4/0 Grit
Supplier: Electron Microscopy Sciences 6211904
Emery paper which is glued to a wooden handle. 11" long x 3/4" wide (279x19.1 mm). Very useful for filing excess resin of sample block tips.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.