Learn More
Electron Microscopy Sciences Flat Emery Stick 4 Grit
Supplier:  Electron Microscopy Sciences 6211940
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Emery paper which is glued to a wooden handle. 11" long x 3/4" wide (279x19.1 mm). Very useful for filing excess resin of sample block tips.
Catalog No. 50-996-763
missing translation for 'QuantityLeft'