Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flavin Adenine Dinucleotide Disodium Salt, ≥95%, MP Biomedicals™

Catalog No. p-4679654 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
250 mg
500 mg
1 g
Cell Biology and Analysis

Chemical Identifiers

CAS 84366-81-4
Molecular Formula C27H31N9Na2O15P2
Molecular Weight (g/mol) 829.52
MDL Number MFCD00151217
InChI Key XLRHXNIVIZZOON-SJFBGTSINA-L
Synonym flavin adenine dinucleotide disodium, unii-67u7uhj04c, flavin adenine dinucleotide disodium salt hydrate, riboflavin 5'-adenosine diphosphate disodium salt, flavin adenine dinucleotide disodium salt, fad-na2, flavine adenine dinucleotide, fad sodium, flavin adenine dinucelotide, fad tn
PubChem CID 131675332
SMILES [Na+].[Na+].CC1=C(C)C=C2N(C[C@H](O)[C@H](O)[C@H](O)COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)N3C=NC4=C(N)N=CN=C34)C3=NC(=O)NC(=O)C3=NC2=C1

Specifications

Color Orange
Assay Percent Range ≥95%
Quantity 250 mg
IUPAC Name disodium;[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4S)-5-(7,8-dimethyl-4-oxido-2-oxobenzo[g]pteridin-10-yl)-2,3,4-trihydroxypentyl] phosphate;hydrate
Formula Weight 829.6
Physical Form Solid
Chemical Name or Material Flavin Adenine Dinucleotide Disodium Salt
EINECSNumber 205-663-1
Recommended Storage Store at 0°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.