Learn More
Hardy Diagnostics™ Flex Minitip Nasopharyngeal (NP) Flock Swab
Shop All Hardy Diagnostics Products
- Individually wrapped, peel pouch
- Nylon flocked tip and a breakpoint at 80 mm
- Bag contains 100 swabs
Specifications
Tip Material
Nylon
Tip Type
Flocked
Sterility
Sterile
Packaging
Individually wrapped, peel pouch
No. per Pack
100
Breakpoint
80 mm
Quantity
100 swabs/bag
Product Type
Swabs and applicators