Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLG Mouse anti-Human, Clone: FLG/1562, Abnova™

Mouse Monoclonal Antibody

Supplier:  Abnova Corporation MAB14528

Catalog No. 89-034-611


missing translation for 'QuantityLeft'
Add to Cart

Description

Description

Mouse monoclonal antibody raised against partial recombinant human FLG.

  • High specificity
  • High reactivity
Specifications

Specifications

FLG
Monoclonal
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
P20930
FLG
Recombinant protein corresponding to amino acids 998-1104 of human FLG.
100 μg
Primary
Human
Liquid
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed)
FLG/1562
Mouse monoclonal antibody raised against partial recombinant human FLG.
In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
ATOD2
Mouse
Protein A/G purification
RUO
2312
Store at 4°C.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

For Research Use Only (RUO)