Learn More
Johannes Lieder Flour and Starch, Spices and Ingredients, Impurities and Adulterations
SCI_ED
7600 Flour and Starch, Spices and Ingredients, Impurities and Adulterations, 25 microscope slides
Supplier:  Johannes Lieder 7600
Catalog No. S02785
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Description
Description
  • 7600 Flour and Starch, Spices and Ingredients, Impurities and Adulterations 
  • 25 Microscope Slides
  • 1. Wheat flour 2. Rye flour 3. Oat meal 4. Potato starch 5. Rice starch 6. Wheat bran 7. Wheat flour adulterated with chalk 8. Rye flour spoiled with moths 9. Corn flour spoiled with spores of corn smut (Ustilago) 10. Spoiled wheat flour (corroded starch) 11. Wheat grain, t.s. for general study 12. Wheat rust (Puccinia), uredinia on wheat leaf t.s. 13. Rye grain, t.s. for general study 14. Mites from meal (Tyroglyphus farinae) 15. Ergot (Claviceps), t.s. of sclerotium 16. Ingredients of rye bread 17. Bacteria from bread, stained 18. Yeast (Saccharomyces), budding cells 19. Fruit rind of lemon, t.s. shows oil glands 20. Milk, stained for fat 21. Almond, t.s. of endosperm 22. Coconut, t.s. of endosperm 23. Cacao powder 24. Cinnamon, ground 25. Aniseed, ground
Specifications
Specifications
General
Cross and Longitudinal Section
Prepared Slide
Elastin Stained Glass
Microscope Slides
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.