Learn More
Cytiva Funnel housing, 150 mL(1/pkg)
Supplier:  Cytiva 4248
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Unique Magnetic Seal Allows Easy, One-Handed Vacuum Filtration of Liquids-No-leak magnetic seal allows one-hand operation. -Polyphenylsulfone construction is compatible with anti-foaming agents and many other solvents.-Convenient. 150 mL size allows for easy fit into small autoclaves and 500 mL size is ideal for filtration of large samples.-Sturdy and safe. Polyphenylsulfone construction provides durability and added safety at a cost less than most glass funnels.-Forceps access point allows easy filter retrieval.-Graduated at 50 mL increments for accurate sample measurement.-New 47 mm Sentino Magnetic Filter Funnels also available.-#8 stopper with proper bore hole size included This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3017
missing translation for 'QuantityLeft'