Learn More
Cytiva Nylaflo™ Nylon Membrane Disc Filter - 0.2 µm, 90 mm (100/pkg)
A Highly Versatile Laboratory Membrane Filter-Excellent chemical compatibility with esters, bases, and alcohols-Naturally hydrophilic-Available in 0.2 and 0.45 µm pore sizes, in diameters ranging from 13 to 142 mm-HPLC certified. Provides assurance that the filter will not add artifacts to your analysis This product is not approved for use in medical, clinical, surgical or other patient applications.