Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluconazole, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. p-7157453 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail

F1355, 86386-73-4, C13H12F2N6O

Spectrum™ Chemical Fluconazole, USP is used to treat and prevent superficial and systematic fungal infections. Known as a triazole antifungal drug, it has a white crystalline appearance and is slightly soluble in water and alcohol. Most commonly found under the trade names Diflucan and Trican and is listed on the World Health Organizations list of essential medicines. Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 86386-73-4
Molecular Formula C13H12F2N6O
Molecular Weight (g/mol) 306.28
InChI Key RFHAOTPXVQNOHP-UHFFFAOYSA-N
IUPAC Name 2-(2,4-difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol
SMILES OC(CN1C=NC=N1)(CN1C=NC=N1)C1=CC=C(F)C=C1F

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 98 to 102%
Grade USP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.