Learn More
Selleck Chemical LLC Fludarabine Phosphate 50mg 75607-67-9 F-ara-A (NSC 312887) Phosphate
Supplier: Selleck Chemical LLC S122950MG
Fludarabine Phosphate (F-ara-A, NSC 312887) is an analogue of adenosine and deoxyadenosine, which is able to compete with dATP for incorporation into DNA and inhibit DNA synthesis.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.