Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flumetasone 98.0+%, TCI America™
SDP

Catalog No. p-7129988 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 2135-17-3
Molecular Formula C22H28F2O5
Molecular Weight (g/mol) 410.458
MDL Number MFCD00056464
InChI Key WXURHACBFYSXBI-GQKYHHCASA-N
PubChem CID 16490
ChEBI CHEBI:34764
IUPAC Name (6S,8S,9R,10S,11S,13S,14S,16R,17R)-6,9-difluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES CC1CC2C3CC(C4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)CO)O)C)O)F)C)F

Specifications

Melting Point 244°C
Color White
Quantity 200 mg
Formula Weight 410.46
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Flumetasone
EINECSNumber (9)-2154
RTECSNumber TU3832000
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.