Learn More
AdipoGen Fluo-3
SDP
Supplier:  AdipoGen CDXF0056M005
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Chemical. CAS 123632-39-3. Formula C36H30Cl2N2O13. MW 769.53. Synthetic. Fluorescent indicator for calcium used in flow cytometry. Fluorescence intensity increases about 40-fold after calcium binding.
Catalog No. 50-256-0907
missing translation for 'QuantityLeft'