Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Isothiocyanate (mixture of 5- and 6- isomers) 97.0+%, TCI America™
SDP

Catalog No. p-7129897 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
100 mg
1 g

Chemical Identifiers

CAS 27072-45-3
Molecular Formula C21H11NO5S
MDL Number MFCD00005063
Synonym Isothiocyanatofluorescein

Specifications

Quantity 100 mg
Formula Weight 389.38
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Fluorescein Isothiocyanate (mixture of 5- and 6- isomers)
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.