Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluoride Standard, 100 ppm F-, Ricca Chemical
BIOPHARMASDP

Catalog No. p-3251693 Shop All Ricca Chemical Company Products
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit
This item is not returnable. View return policy
MTO (Made-To-Order) item. This item is subject to the following cancellation policy: This item may be cancelled within 10 days of order placement otherwise is not returnable for credit

Chemical Identifiers

CAS 7681-49-4, 7732-18-5
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Packaging Natural Poly Bottle
Quantity 500 mL
Color Colorless
Concentration or Composition Notes 100ppm F-
Equivalence 1mL = 0.1mg F-
Physical Form Liquid
Chemical Name or Material Fluoride Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.