Learn More
CTL Scientific Supply Corp FLUORIDE TEST,30 TEST DISKS
Supplier: CTL Scientific Supply Corp 90734
Fluoride Test - 30 discs and reagent for the semi-quantitative determination of fluoride ions. Gradation of scale: 0 - 2 - 5 - 10 - 20 - 50 - 100 mg/l F- (each kit contains a color comparison scale)
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.