Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fluphenazine, 98+%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
Melting Point | 268°C to 274°C |
Color | Brown to Yellow |
Quantity | 1 g |
UN Number | UN2811 |
Merck Index | 14,4189 |
Formula Weight | 437.52 |
Percent Purity | ≥98% |
Physical Form | Powder |
Chemical Name or Material | Fluphenazine |
Safety and Handling
Hazard Category | Danger |
Hazard Statement | GHS H Statement H301 Toxic if swallowed. |
Precautionary Statement | GHS P Statement P264-P270-P301+P310-P321-P405-P501a Wash thoroughly after handling. Do not eat, drink or smoke when using this product. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Specific treatment (see label). Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
RTECSNumber | TL9730000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction