Learn More
TARGETMOL CHEMICALS INC FLUPHENAZINE DECANOATE 100MG
Supplier:  TARGETMOL CHEMICALS INC T7943
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Fluphenazine decanoate is dopamine D2 receptor blockeand is a long-acting phenothiazine neuroleptic that used to treat schizophrenia. purity: 100%
Catalog No. 50-257-9106
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.