Learn More
TARGETMOL CHEMICALS INC FLUPIRTINE MALEATE 200MG
Supplier:  TARGETMOL CHEMICALS INC T6504
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Also available in 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Flupirtine maleate (Katadolon maleate) a centrally acting non-opioid analgesia is the salt form of Flupirtine. It is an NMDA receptor antagonist and also a specific neuronal potassium channel opener. purity: 99%
Catalog No. 50-257-6500