Learn More
Chemglass Life Sciences Funnel, Weighing, Disposable PP, 20mL Solid Capacity, 10mL Liquid Capacity
SDP
Supplier:  Chemglass Life Sciences CG176018
Encompass_Preferred
Funnel, Weighing, Disposable PP, 20mL Solid Capacity, 10mL Liquid Capacity.• Available in anti-static PP, PP and borosilicate glass• With flat bottoms and designed to fit narrow neck vessels
Catalog No. 50-872-125
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.