Learn More
Sklar Instruments Foerster Sponge Forceps, Sklar 12 in., Straight, Serrated, Stainless Steel. EA.
Supplier: Sklar Instruments 872490
Foerster Sponge Forceps, Sklar. 12 in., Straight, Serrated, Stainless Steel. Sklar Foerster Sponge Forceps, which may also be known as Foerster-Ballenger Sponge Forceps, are commonly used in surgical procedures to hold gauze squares that will absorb excess fluids and blood from the surgical site or hemostasis. The ratcheted locking mechanism of these finger ring forceps securely grip the gauze and hold it in place. Foerster Sponge Forceps that are longer in length are often used in bariatric surgeries and the curved version may be used for removing the placenta in OB/GYN procedures. This product is straight with oval, fenestrated, serrated tips and a length of 12 inches.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.