Learn More
Memmert Memmert Universal Heating and Drying Laboratory Oven UF30. 30L - 230 Volt, Forced-Air Convection, Stainless Steel Construction with SingleDISPLAY digital controller.
SDP
Supplier:  Memmert UF30 230V
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Memmert Universal heating and drying Oven - this model is the 230 volt, forced-air convection, with SingleDISPLAY digital controller. Interior dimensions 400/320/211-W/H/D mm. Includes (1) Stainless Steel Grid Shelf, Quick Start Guide, Calibration Certificate and Power Cord - Working Range 10C above ambient to +300C.
Catalog No. 50-225-4274
missing translation for 'QuantityLeft'