Learn More
Electron Microscopy Sciences Forensic Field Sampler Stub without Adhesive Carbon Tab 8 mm
Supplier: Electron Microscopy Sciences 7648031
Glass specimen vial has a high-purity certified 12.7 mm SEM pin stub mounted into the plastic cap. It is ideal for collecting forensic evidence with minimum interference and/or contamination from the sampler. The specimen arrives at the lab ready for carbon coating for SEM/EDX investigation. The Forensic Field Sampler can be securely stored in the glass specimen vial and easily shipped with the cap holding the sampler stub in place. Vial is 25 mm OD x 55 mm high. .
Pin length: 8 mm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.