Learn More
Formaldehyde 37%, Vion Biosciences
Shop All Aldon Products- Stabilize the fine structural details of tissue samples with this histology-grade formaldehyde fixative.
Specifications
Boiling Point
96°C (205°F)
Closure Type
Screw Cap
Color
Colorless
Content And Storage
Store in a cool, well-ventilated area away from incompatible substances. Keep away from ignition sources.
Packaging Type
F Style UN Bottle
Concentration
37%
For Use With (Equipment)
Microscope
Grade
Histological
Product Line
Vion Histology
Product Type
Fixative
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.