Learn More
NEOGEN FORMALDEHYDE ASSAY KIT
SDP
Supplier:  NEOGEN 700004284
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass
Formaldehyde Assay Kit50 assays (manual) / 500 assays (microplate) / 500 assays (auto-analyser)Extended cofactors stability. Dissolved cofactors stable for > 1 year at 4°C.All reagents stable for > 2 yearsSpecific for formaldehydeSuitable for manual and auto-analyser formats Suitable for manual, microplate and auto-analyser formatsStandard includedMega-Calc™ software tool is available from our website for hassle-free raw data processing
Catalog No. NC2173712