Learn More
Electron Microscopy Sciences Formaldehyde/Zinc Fixative 1 LT
Supplier:  Electron Microscopy Sciences 1567501
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
Prepared: Ready-to-use. Preserves fine nuclear details and good for immunostaining. Contains no mercury. Can be used with automated tissue processors. Works well with standard and special stains because zinc is a stain enhancing mordant.
Catalog No. 50-980-478