Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Formalin Dogfish Sharks
SDPSCI_ED

For use in exploring anatomy of cartilaginous fish. Carolina™ Formalin Dogfish Sharks are ideal for comparative and vertebrate anatomy classes.

Supplier:  Carolina™ S226811

 View more versions of this product

Catalog No. S37636



missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart

Description

Description

  • Member of class Chondrichthyes
  • Mature specimens
  • Characteristics: cartilaginous skeleton, five pairs of gill slits, no swim bladder or operculum
  • Shipped in Carosafe™, Carolina's proprietary shipping and holding fluid
Specifications

Specifications

Double
55.8 to 68.5cm
Formalin
1/bag
22 to 27 in.
Bag
Preserved specimen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.