Learn More
Electron Microscopy Sciences NeutraGreen™ Neutralizer 2.5 GL
Supplier: Electron Microscopy Sciences 2414602
Neutralizer that transforms waste formalin into an odorless, 100 percent nonhazardous material. Comes in liquid form with no powders to mix or dissolve. One of the fastest neutralizers available with a reaction time of only 2-3 hours. Third-party environmental data confirms NeutraGreen will neutralize waste formalin to a material that is non-toxic as well as non-hazardous.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.