Learn More
Electron Microscopy Sciences F-gard-ER Formalin Spill Control 11 OZ
Supplier:  Electron Microscopy Sciences 2414006
Encompass_Preferred
For the handling and disposing of formaldehyde containing solutions. It immobilizes, cools, and contains spills. As well, it reduces the escape of formaldehyde gas and significantly reduces formalin concentration. F-gard- ER aids in reducing exposure to formaldehyde gases by immobilizing the formalin and transforming it to an easily removed semi-solid mass. The mass cools rapidly minimizing formaldehyde off-gassing.
Catalog No. 50-260-16
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.