Learn More
Ted Pella Inc FORMVAR 400 MESH COPPER 01702F
Formvar, 400 mesh, copper, pkg/50 Substrates, Support Films for Transmission Electron Microscopy Grids Formvar: A film of pure Formvar, with no stabilizing coating. These films are useful for your customized coating of stabilizing material or for the support of thin sections.