Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on Gilder F2 Finder Gold Grid
Supplier:  Electron Microscopy Sciences FF200F2AU50
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a Gilder Finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Gold
Catalog No. 50-190-0644
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.