Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on Square 400 mesh Copper Grid
Support Film on grids has become a main product line for us since the demand for high quality coated grids has increased. To make your microscopy work easier and to save you a great deal of time we offer you a complete line. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper