Learn More
Electron Microscopy Sciences Formvar Support Film 5-6 nm thick on London Finder LF135 Copper Grid
Supplier:  Electron Microscopy Sciences FFLF135CU25
Encompass Procurement Services
Non-distribution item offered as a customer accommodation; additional freight charges may apply.
Learn More
Encompass_Preferred
A thin film of pure formvar resin coats the shiny side of a London finder grid. Coated grids are optically checked and batch tested in the electron microscope. Packed in grid storage box. Standard coating is 5 to 6 nm thick.
Grid: Copper
Catalog No. 50-190-0655