Learn More
MAC-MOD Analytical Fortis Direct Guard Tips (5/pk)
SDP
Supplier:  MAC-MOD Analytical DCTIP5
Encompass_Preferred
Fortis HPLC and UHPLC columns are packed with 100 A high-surface area silica-based columns designed with optimized bonding chemistries for uniform ligand coverage. This proprietary process improves method development and improves LCMS sensitivity by eluting compounds in higher organic mobile phases, as well as when scaling to preparative separation elutions in higher organic leads to quicker “dry-down” times. C18, C8, Diphenyl, H2o, Cyano and Amino bonded phases are all available on 1.7, 3, 5 and 10 um particle sizes. HILIC and DIOL phases are also available in the Fortis HILIC range. Fortis columns are available in a wide range of column IDs and column lengths including 75 micron, 200 micron, 2.1, 3.0, 4.6, 10 and 21.2 mm ID columns other column IDs and lengths available upon request. Guard columns are available and require guard holder DCGUA1.
Catalog No. 50-234-0052
missing translation for 'QuantityLeft'
Explore all special offers
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.