Learn More
Foss™ Exhaust Manifold for Digestors
Facilitate fume removal and containment
Supplier: Foss™ 60062243
Description
Ensures Good Laboratory Practice (GLP).
- Strongly recommended for use with all digestion procedures
- Should be connected to water aspirator (supplied) or to a suitable scrubber unit
Specifications
Exhaust Manifold TC2508 Series | |
For use with all digestion procedures | |
Digestion unit | |
1 Ea. |