Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foss™ Kjeltec™ 8100 Manual Distillation Unit

A simple and reliable means to safe, manual distillations

Supplier:  Foss™ 60063319

Catalog No. TC8100001

Alert:

This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.


Includes: Tanks with level sensors for alkali/water/waste; Receiver flask; 100 and 250mL tubes; Combi-adapter for 100, 250, 400 and 750mL tubes.

Description

Description

  • Automatic dilution, alkali addition, distillation and tube emptying for ease of use
  • Variable output steam generator broadens application area to other volatile components
  • Official and accurate procedures (e.g., ISO, AOAC, EPA, DIN) simplify validation
  • Patented SAfE™ feature for safer distillation of tubes with salt cakes
  • Built-in safety systems for user protection
  • Self-adjusting cooling water control saves water and reduces costs
  • Bellows pumps for accurate reagent dispensing
  • Alkali-resistant plastic splash head for long lifetime

Order Info

Dimensions: 48L x 58W x 69cm H. Net weight, 66 lb. (30kg).

Specifications

Specifications

200/240V 50/60Hz
Tanks with level sensors for alkali/water/waste; receiver flask; 100 and 250ml tubes; combi-adapter for 100, 250, 400 and 750ml tubes
Automatic dilution, alkali addition, distillation and tube emptying
48cm
27.16 in.
66 lb.
18.8 in.
200/240V
69cm
50/60Hz
Manual Distillation Unit
30kg
22.8 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.