Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-Curve™ Four Compartment Glove Dispenser with Individual Lids, Front Access

Catalog No. p-7235469 Shop All S-Curve Technologies Products
Click to view available options
Material:
Acrylic
PETG
This item is not returnable. View return policy
This item is not returnable. View return policy

Four compartment dispenser includes four individually opening lids, with front access holes for easy product dispensing.

  • Four compartment dispenser with individually opening lids
  • Front access holes for easy product dispensing
  • Can be mounted with heavy-duty wall brackets (included)
  • Flame treated edges for a smooth finish

Specifications

Product Type Glove Dispenser
Color Clear
Dimensions (D x W x H) Interior 11.5 x 20 x 12 in.
Holds Gloves
Material Acrylic
Mounting Keyhole Slots
No. of Compartments 4
Product Style Top Loading
Includes Heavy-duty wall mounting bracket
Thickness (English) 0.25 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.